Anti-CYP2B6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-2352R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily B, Polypeptide 6
- Antigen symbol:CYP2B6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1555
- Antigen synonyms:CYP2B7|CPB6|P450|CYP2B7P|IIB1|CYPIIB6|CYP2B|EFVM
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:397-431/491
- Purification:Purified by Protein A
- Pk:100 µl
Cytochromes P45 are a group of heme-thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH-dependent electron transport pathway. It oxidizes a variety of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. Acts as a 1,4-cineole 2-exo-monooxygenase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CYP2B6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity: