Order Entry
Germany
ContactUsLinkComponent
 
Anti-MMP16 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-1856R-CY7
 :  Bioss
Anti-MMP16 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-1856R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 16
  • Antigen symbol:
    MMP16
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    4325
  • Antigen synonyms:
    DKFZp761D112|MT-MMP2|MMP-X2|MT-MMP3|MT3-MMP|C8orf57
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    500-550/607
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Endopeptidase that degrades various components of the extracellular matrix, such as collagen type III and fibronectin. Activates progelatinase A. Involved in the matrix remodeling of blood vessels. Isoform short cleaves fibronectin and also collagen type III, but at lower rate. It has no effect on type I, II, IV and V collagen. However, upon interaction with CSPG4, it may be involved in degradation and invasion of type I collagen by melanoma cells.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP16
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat