Anti-GADD45A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-1360R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Growth arrest And DNA-damage-inducible protein GADD45 alpha
- Antigen symbol:GADD45A
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1647
- Antigen synonyms:GADD45A|DDIT-1|DNA-damage-inducible transcript 1|GADD45|Growth arrest and DNA-damage-inducible protein GADD45 alpha|DDIT1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:100-150/165
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
In T-cells, functions as a regulator of p38 MAPKs by inhibiting p88 phosphorylation and activity (By similarity). Might affect PCNA interaction with some CDK (cell division protein kinase) complexes; stimulates DNA excision repair in vitro and inhibits entry of cells into S phase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GADD45
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat