Order Entry
Germany
ContactUsLinkComponent
 
Anti-GADD45A Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1360R-CY5
Supplier:  Bioss
Anti-GADD45A Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1360R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Growth arrest And DNA-damage-inducible protein GADD45 alpha
  • Antigen symbol:
    GADD45A
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1647
  • Antigen synonyms:
    GADD45A|DDIT-1|DNA-damage-inducible transcript 1|GADD45|Growth arrest and DNA-damage-inducible protein GADD45 alpha|DDIT1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    100-150/165
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

In T-cells, functions as a regulator of p38 MAPKs by inhibiting p88 phosphorylation and activity (By similarity). Might affect PCNA interaction with some CDK (cell division protein kinase) complexes; stimulates DNA excision repair in vitro and inhibits entry of cells into S phase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GADD45
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat