Specifications
- Antibody type:Primary
- Antigen name:Glutathione S-transferase Theta 2
- Antigen symbol:GSTT2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2953
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Glutathione (GSH) is a tripeptide antioxidant which reduces disulfide bonds between cytoplasmic proteins. The constitutive enzyme glutathione reductase transforms glutathione into its reduced state which ultimately can provide a measure of cellular toxicity. GSTT2 (glutathione S-transferase theta-2), also known as GST class-theta-2, is a 244 amino acid enzyme with sulfatase activity that functions in conjugating reduced glutathione to hydrophobic electrophiles. GSTT2 exists as a homodimer in the cytoplasm and is expressed in low levels in the liver and the lung. GSTT2 belongs to the GST superfamily and contains both a GST C-terminal and a GST N-terminal domain. The gene encoding GSTT2 exists on human chromosome 22.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GSTT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat