Anti-TUBG1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1322R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tubulin, Gamma 1
- Antigen symbol:TUBG1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7283
- Antigen synonyms:TUBGCP1|CDCBM4|TUBG|GCP-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:415-450/451
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Tubulin is the major constituent of microtubules. The gamma chain is found at microtubule organizing centers (MTOC) such as the spindle poles or the centrosome. Pericentriolar matrix component that regulates alpha/beta chain minus-end nucleation, centrosome duplication and spindle formation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TUBG1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat