Anti-KDSR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13233R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:3-Ketodihydrosphingosine Reductase
- Antigen symbol:KDSR
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2531
- Antigen synonyms:SDR35C1|FVT1|DHSR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Catalyzes the reduction of 3-ketodihydrosphingosine (KDS) to dihydrosphingosine (DHS).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KDSR
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat