Anti-ERCC8 Rabbit Polyclonal Antibody (Cy5®)
Product Details & Documents
Substrate-recognition component of the CSA complex, a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex, involved in transcription-coupled nucleotide excision repair. The CSA complex (DCX(ERCC8) complex) promotes the ubiquitination and subsequent proteasomal degradation of ERCC6 in a UV-dependent manner; ERCC6 degradation is essential for the recovery of RNA synthesis after transcription-coupled repair. It is required for the recruitment of XAB2, HMGN1 and TCEA1/TFIIS to a transcription-coupled repair complex which removes RNA polymerase II-blocking lesions from the transcribed strand of active genes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ERCC8
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-13095R-CY5
Datasheet
Specifications
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Store at 4°C for 12 months
- Host:Rabbit
- Concentration:1 μg/μl
- Cross adsorption:No
- Shipping temperature:4°C
- Antigen name:Excision Repair Cross-Complementation Group 8
- Cat. No.:BOSSBS-13095R-CY5
- Reactivity:Human|Rat|Mouse
- Gene ID:1161
- Antigen synonyms:CSA|UVSS2|CKN1
- Immunogen:180-230/396
- ImmunoChemistry:Yes
- Conjugation:Cy5®
- Antigen symbol:ERCC8
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



