Anti-CD28 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-1297R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CD28 Molecule
- Antigen symbol:CD28
- Clonality:Polyclonal
- Conjugation:Cy3®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:12487
- Antigen synonyms:Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:20-45/218
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in T-cell activation, the induction of cell proliferation and cytokine production and promotion of T-cell survival.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD28
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat