Anti-DOCK2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-12431R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:dedicator of cytokinesis 2
- Antigen symbol:DOCK2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1794
- Antigen synonyms:IMD40
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:500-550/1830
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in cytoskeletal rearrangements required for lymphocyte migration in response of chemokines. Activates RAC1 and RAC2, but not CDC42, by functioning as a guanine nucleotide exchange factor (GEF), which exchanges bound GDP for free GTP. May also participate in IL2 transcriptional activation via the activation of RAC2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DOCK2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat