- Antibody type:Primary
- Antigen name:dedicator of cytokinesis 4
- Antigen symbol:DOCK4
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:9732
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
DOCK 4 is a cytoplasmic peripheral membrane protein that belongs to the DOCK family of cytokinesis-regulating proteins. Expressed ubiquitously with highest expression in prostate, ovary and skeletal muscle, DOCK 4 functions as a guanine nucleotide exchange factor (GEF) that activates the small GTPase Rap 1 and, via this activation, plays a role in the regulation of adherens junctions between cells. Similar to other DOCK family members, DOCK 4 contains an N-terminal SH3 domain, a C-terminal proline-rich region and two internal DOCK homology regions designated DHR1 and DHR2. Defects in the gene encoding DOCK 4 result in the inactivation of Rap 1 and are, thus, implicated in the pathogenesis of various cancers such as ovarian, prostate, glioma and colorectal carcinomas. Four isoforms of DOCK 4 are expressed due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DOCK4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat