Anti-DOCK7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-11825R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:dedicator of cytokinesis 7
- Antigen symbol:DOCK7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ZIR2|EIEE23
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
DOCK 7 is a 2,140 amino acid protein that localizes to developing axons and contains one DHR-1 domain and one DHR-2 domain. Expressed in a variety of tissues, DOCK 7 functions as a guanine nucleotide exchange factor (GEF) that specifically activates Rac 1 and Rac 3 by catalyzing the exchange of bound GDP for free GTP. Multiple isoforms of DOCK 7 exist due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DOCK7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat