Anti-FGGY Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-11689R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:FGGY carbohydrate kinase Domain containing
- Antigen symbol:FGGY
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55277
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:175-225/551
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a protein that phosphorylates carbohydrates such as ribulose, ribitol, and L-arabinitol. Genome-wide association studies in some populations have found an association between polymorphisms in this gene and sporadic amyotrophic lateral sclerosis, but studies of other populations have not been able to replicate this association. Alternative splicing results in multiple transcript variants. [provided by RefSeq, May 2013]
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FGGY
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat