- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase 1
- Antigen symbol:CDK1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:12534
- Antigen synonyms:p34 protein kinase|cell division control protein 2 homolog|P34CDC2|Cell Division Cycle 2|CDC2|CDC28A|G1 to S and G2 to M|cell cycle controller CDC2|cell division protein kinase 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:250-297/297
- Purification:Purified by Protein A
- Pk:100 µl
The cell division control protein cdc2, also known as cyclin dependent kinase 1 (Cdk1) or p34/cdk1, plays a key role in the control of the eukaryotic cell cycle, where it is required for entry into S phase and mitosis. Cdc2 exists as a complex with both cyclin A and cyclin B. The best characterized of these associations is the Cdc2 p34 cyclin B complex, which is required for the G2 to M phase transition. Activation of Cdc2 is controlled at several steps including cyclin binding and phosphorylation of threonine 161. However, the critical regulatory step in activating cdc2 during progression into mitosis appears to be dephosphorylation of Tyr15 and Tyr14. Phosphorylation at Tyr15 and inhibition of Cdc2 is carried out by WEE1 and MIK protein kinases while Tyr15 dephosphorylation and activation of Cdc2 is carried out by the cdc25 phosphatase. The isoform CDC2deltaT is found in breast cancer tissues. Furthermore, cdc2/Cdk1 is a key mediator of neuronal cell death in brain development and degeneration.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDK1
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: