Anti-PPARD Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-0250R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Peroxisome Proliferator-Activated Receptor Delta
- Antigen symbol:PPARD
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:19015
- Antigen synonyms:Peroxisome Proliferator-Activated Nuclear Receptor |Pparb/d|NUC1|PPAR delta|PPAR-beta|Nuclear Hormone Receptor 1|Pparb|Nuclear Receptor Subfamily 1 Group C Member 2|PPAR-delta|PPAR[b]|NUC-1|Nr1c2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1-50/440
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Ligand-activated transcription factor. Receptor that binds peroxisome proliferators such as hypolipidemic drugs and fatty acids. Has a preference for poly-unsaturated fatty acids, such as gamma-linoleic acid and eicosapentanoic acid. Once activated by a ligand, the receptor binds to promoter elements of target genes. Regulates the peroxisomal beta-oxidation pathway of fatty acids. Functions as transcription activator for the acyl-CoA oxidase gene. Decreases expression of NPC1L1 once activated by a ligand (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PPARD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat