Anti-EPHB2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-0247R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:EPH Receptor B2
- Antigen symbol:EPHB2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:2048
- Antigen synonyms:EK5|Hek5|CAPB|ERK|EPHT3|PCBC|Tyro5|DRT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Eph receptors play important roles in axon guidance at the midline. In the auditory system, growth of axons across the midline is an important determinant of auditory function. The avian cochlear nucleus, n. magnocellularis (NM), makes bilateral projections to its target, n. laminaris (NL).
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: EPHB2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat