Zinc hydroxide, white ≥99.5%, powder
Catalog # ICNA0521996980
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identity:Passes test
- FTIR:Conforms to Standard
- Iron:0,005%
- Lead:0,0012%
- Chloride:0,006%
- Magnesium & alkaline earths:0,25%
- Assay:99,5%
- HCI insoluble substances:0,02%
Specification Test Results
About this item
:
:99.40 g/mol
Melting Pt:125 °C (lit.)
Density:∼3.053 g/cm³ (20 °C) (lit.)
UN:3077
ADR:9,III
CAS Number:20427-58-1
Formula:H₂O₂Zn
MDL Number:MFCD00049620