Microbiology manifold
Catalog # 516-1038
Supplier: Cytiva (Formerly Pall Lab)
Specifications
- Description:3-place manifold base
- Seq:01
- Pk:1
Specifications
About this item
This microbiology manifold, with modular design, allows you to optimise testing without sacrificing cleanliness.
- Can be customised to fit workflow, test area and autoclave
- Slip-fit adapters enable the use of any Pall microbiology device on the same manifold
- Right- or left-handed operation
With a 3-place manifold and a coupling device, an additional manifold can be added to make a 6-place manifold. A preventative maintenance schedule can be designed without delays to critical testing.
Adapters are required and need to be ordered separately.
MicroFunnel™ adapter: Ideal for MicroFunnel™, MicroFunnel™ ST, MicroFunnel™ Plus and MicroCheck® devices.
Sentino® funnel adapter: Ideal for Sentino® filter funnels.
Standard adapter: Ideal for Sentino® magnetic filter funnels.
Elongated standard adapter: Ideal for magnetic filter funnels.
Ordering information: The microbiology manifold is supplied as a set and comes with 3-place manifold base, 3 manifold valves, 1 end cap and 1 hose barb cap. Different adapters and coupling devices are available. Adapters are required and need to be ordered separately.