Cut resistant gloves, Vertigo White PU 3
Supplier: HONEYWELL SAFETY
Certificates
About this item
These white Spectra®/Lycra® knitted gloves feature a white PU coating on the palm and fingers tips. The Spectra®/Lycra® thread provides outstanding properties against cutting while the PU coating provides outstanding abrasion resistance.
- Knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- Maximum comfort and added sturdiness
- Cool Spectra® touch gives an added sensation of comfort
- Ventilated back optimises breathability
- White colour enhances hygiene and cleanliness properties
Ideal for fine handlings in a dry environment. Typical applications include: Automotive and part manufacturing, aviation, building and construction, catering, energy or electricity, glass, metal and paper industries, logistics, maintenance, Telecoms and transportation.
Certifications: EN 388:2016 (4X42B); Cat. II.
Caution: Do not expose this glove to the heat.
Specifications
- Catalog No:
- DALL2318769-06
- DALL2318769-10
- DALL2318769-09
- DALL2318769-08
- DALL2318769-07
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Spectra®/Lycra®
- Spectra®/Lycra®
- Spectra®/Lycra®
- Spectra®/Lycra®
- Spectra®/Lycra®
- Finish:
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)