Sealing rings for dessicators
Catalog # 467-2240
Supplier: KLEINFELD
Specifications
- Ø:100 mm
- For:Dessicators
- Description:Sealing ring
- Seq:01
- Pk:1
Specifications
About this item
Soft rubber sealing rings, makes grease unnecessary. Suitable for desiccators and Witt jars.