ContactUsLinkComponent
Briggs and Salt Mixture, MP Biomedicals
Briggs and Salt Mixture, MP Biomedicals
Catalog # ICNA0290283410
Supplier:  MP Biomedicals
undefined
Briggs and Salt Mixture, MP Biomedicals
Catalog # ICNA0290283410
Supplier:  MP Biomedicals
Specifications
  • Description:
    Briggs and salt mixture
  • Pk:
    10 kg
Specifications
About this item
This powdered salt mixture is used as animal diet.
It contains calcium carbonate, calcium phosphate, copper sulphate, ferric citrate, magnesium sulphate, manganese sulphate, potassium chloride, potassium iodate, sodium chloride, sodium phosphate dibasic and zinc carbonate.
Caution: Store at room temperature.