


Phenolphthalein solution 10 g/l in ethanol 96% (v/v) Reag. Ph. Eur. R1 indicator for titration
Catalog # KRAF26295.2700
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:318.33 g/mol
UN:1170
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913