Buffer indicator tablets for water hardness
KRAF20506.3601
:
- Pk:1.000
- Description:Buffer indicator solution
Indicator buffer tablets used for the determination of the water hardness with EDTA.
Indicator buffer tablets used for the determination of the water hardness with EDTA.