Order Entry
Germany
ContactUsLinkComponent
Ammonium hydrogen carbonate, LiChropur™ for HPLC, Supelco®
Irritant
Ammonium hydrogen carbonate, LiChropur™ for HPLC, Supelco®
Supplier:  Merck
Certificates
Irritant
Ammonium hydrogen carbonate, LiChropur™ for HPLC, Supelco®
Supplier:  Merck

Specification Test Results

  • Assay (acidimetric):
    99,0 - 101,0%
  • Identity:
    Passes test
  • Non-volatile matter:
    ≤0,05%
  • Aluminium:
    ≤2 ppm
  • Calcium:
    ≤5 ppm
  • Copper:
    ≤0,1 ppm
  • Iron:
    ≤5 ppm
  • Potassium:
    ≤1 ppm
  • Magnesium:
    ≤10 ppm
  • Sodium:
    ≤1 ppm
  • HPLC gradient suitability test for chromophoric impurities based on 4'-hydroxyacetophenone:
    ≤10 ppm
  • Chromophore purity:
    ≥99,999%

Specification Test Results