Order Entry
Germany
ContactUsLinkComponent
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ICNA0855526
Supplier:  MP Biomedicals
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ICNA0855526
Supplier:  MP Biomedicals

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulins
  • Antigen symbol:
    Ig
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Antibody fragment:
    F(ab')2
  • Concentration:
    Total Protein: 1.05 mg/mL; F/P Ratio: 5.2 mol/mol
  • Shipping temperature:
    RT
  • Purification:
    Antibody and FITC are conjugated under defined conditions to obtain the specified molar ratio (3-6 mol/mol). Conjugated protein is purified by gel filtration. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide and 10% glycerol (v/v), adjusted to standard titer, filtered through a 0.22 µm filter, and vialed.
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

This product is fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat affinity purified F(ab')2 fragments to mouse immunoglobulins (IgG,IgA,IgM) and buffer salts.The total protein is measured by absorbance (A)280 nm); F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the affinity purified F(ab')2fragments is standardized with an in-house control by immunoelectrophoresis. The affinity purified F(ab')2 fragment is tested for purity at 10 mg/mL using immunoelectrophoresis. The product is goat F(ab')2; no trace of albumin, whole molecule IgG or Fc is detected. This product contains sodium azide. To prevent formation of toxic vapors, do not mix with strongly acidic solutions. To prevent formation of potentially explosive metallic azides in metal plumbing, always wash into drain with copious quantities of water.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: F(ab')2
Reactivity: Mouse