Anti-IgM Goat Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ICNA0855153
Supplier: MP Biomedicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Lyophilized
- Concentration:Total Protein: 8.0 mg/mL; Antibody Protein: 3.5 mg/mL
- Shipping temperature:RT
- Purification:Antibody and FITC are conjugated under defined conditions to obtain the specified molar ratio (3-6 mol/mol). Conjugated protein is purified by gel filtration. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed, and lyophilized.
- Size:2 ml
- Pk:2 mL
Specifications
About this item
This product is the lyophilized powder of fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat IgG fraction to human IgM (5Fc µ) and buffer salts.The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized by equivalence-point precipitation. The IgG fraction is tested for purity and specificity at 40 mg/mL using immunoelectrophoresis. The product is mostly goat IgG; no trace of albumin is detected.
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human