Deep well plates, 96-well, DispoDialyzer™
The single use 96-well DispoDialyzer™ is ideal for the simultaneous preparation of 96 samples.
- Each well in this system has a separate membrane, thus eliminating the possibility of sample contamination and leakage
- Very high reproducibility across different wells
- Excellent sample recovery
- Sealed with twelve 8-cap strips