Order Entry
Germany
ContactUsLinkComponent
 
Anti-RASA3 Rabbit Polyclonal Antibody
  BWRLBS5730
Anti-RASA3 Rabbit Polyclonal Antibody
  BWRLBS5730

 

  • Antibody type:
    Primary
  • Antigen name:
    RAS p21 protein activator 3
  • Antigen symbol:
    RASA3
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 290-335 of Human RASA3.

RASA3 pAb detects endogenous levels of RASA3 protein.

Type: Primary
Antigen: RASA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat