Order Entry
Germany
ContactUsLinkComponent
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]
Catalog # BWRLBS5058
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]
Catalog # BWRLBS5058

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-Cdk1/Cdc2
  • Clonality:
    Polyclonal
  • Clone:
    Y15
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.

p-Cdk1/Cdc2 (Y15) pAb detects endogenous levels of Cdk1/Cdc2 protein only when phosphorylated at Tyr15.

Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: Y15
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat