Order Entry
Germany
ContactUsLinkComponent
Anti-p-FGFR1 Rabbit Polyclonal Antibody [clone: Y654]
Anti-p-FGFR1 Rabbit Polyclonal Antibody [clone: Y654]
Catalog # BWRLBS4686
Anti-p-FGFR1 Rabbit Polyclonal Antibody [clone: Y654]
Catalog # BWRLBS4686

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-FGFR1
  • Clonality:
    Polyclonal
  • Clone:
    Y654
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human FGFR1 around the phosphorylation site of Tyrosine 654.

p-FGFR1 (Y654) pAb detects endogenous levels of FGFR1 protein only when phosphorylated at Tyr654.

Type: Primary
Antigen: p-FGFR1
Clonality: Polyclonal
Clone: Y654
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat