Anti-RTCD1 Rabbit Polyclonal Antibody
BWRLBS5918
- Antibody type:Primary
- Antigen symbol:RTCD1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 321-366 of Human RTCD1.
RTCD1 pAb detects endogenous levels of RTCD1 protein.
Type: Primary
Antigen: RTCD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat