Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T14]
Catalog # BWRLBS4265
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Cdk1/Cdc2
- Clonality:Polyclonal
- Clone:T14
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Threonine 14.
p-Cdk1/Cdc2 (T14) pAb detects endogenous levels of Cdk1/Cdc2 protein only when phosphorylated at Thr14.
Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: T14
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat