Anti-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: N133]
Catalog # BWRLBS1914
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cdk1/Cdc2
- Clonality:Polyclonal
- Clone:N133
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 771-820 of Human Cdk1/Cdc2.
Cdk1/Cdc2 (N133) pAb detects endogenous levels of Cdk1/Cdc2 protein.
Type: Primary
Antigen: Cdk1/Cdc2
Clonality: Polyclonal
Clone: N133
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat