Anti-STAC2 Rabbit Polyclonal Antibody [clone: R248]
Catalog # BWRLBS3398
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:STAC2
- Clonality:Polyclonal
- Clone:R248
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 212-270 of Human STAC2.
STAC2 (R248) pAb detects endogenous levels of STAC2 protein.
Type: Primary
Antigen: STAC2
Clonality: Polyclonal
Clone: R248
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat