Anti-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: E8]
BWRLBS1820
- Antibody type:Primary
- Antigen symbol:Cdk1/Cdc2
- Clonality:Polyclonal
- Clone:E8
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 771-820 of Human Cdk1/Cdc2.
Cdk1/Cdc2 (E8) pAb detects endogenous levels of Cdk1/Cdc2 protein.
Type: Primary
Antigen: Cdk1/Cdc2
Clonality: Polyclonal
Clone: E8
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat