Anti-Vimentin Rabbit Polyclonal Antibody [clone: S55]
BWRLBS3182
- Antibody type:Primary
- Antigen symbol:Vimentin
- Clonality:Polyclonal
- Clone:S55
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 111-60 of Human Vimentin.
Vimentin (S55) pAb detects endogenous levels of Vimentin protein.
Type: Primary
Antigen: Vimentin
Clonality: Polyclonal
Clone: S55
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human