Anti-Adducin beta Rabbit Polyclonal Antibody [clone: E497]
BWRLBS2523
- Antibody type:Primary
- Antigen symbol:Adducin beta
- Clonality:Polyclonal
- Clone:E497
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 470-520 of Human Adducin beta.
Adducin ? (E497) pAb detects endogenous levels of Adducin ? protein.
Type: Primary
Antigen: Adducin beta
Clonality: Polyclonal
Clone: E497
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat