Anti-DUSP19 Rabbit Polyclonal Antibody [clone: F133]
Catalog # BWRLBS2336
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:DUSP19
- Clonality:Polyclonal
- Clone:F133
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 100-150 of Human DUSP19.
DUSP19 (F133) pAb detects endogenous levels of DUSP19 protein.
Type: Primary
Antigen: DUSP19
Clonality: Polyclonal
Clone: F133
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat