Order Entry
Germany
ContactUsLinkComponent
Anti-C/EBP zeta Rabbit Polyclonal Antibody [clone: D24]
Anti-C/EBP zeta Rabbit Polyclonal Antibody [clone: D24]
Catalog # BWRLBS2297
Anti-C/EBP zeta Rabbit Polyclonal Antibody [clone: D24]
Catalog # BWRLBS2297

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    C/EBP zeta
  • Clonality:
    Polyclonal
  • Clone:
    D24
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to the N-terminus of Human C/EBP zeta.

C/EBP ? (D24) pAb detects endogenous levels of C/EBP ? protein.

Type: Primary
Antigen: C/EBP zeta
Clonality: Polyclonal
Clone: D24
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human