Order Entry
Germany
ContactUsLinkComponent
Anti-p-GABAA Rbeta1 Rabbit Polyclonal Antibody [clone: S434]
Anti-p-GABAA Rbeta1 Rabbit Polyclonal Antibody [clone: S434]
Catalog # BWRLBS4078
Anti-p-GABAA Rbeta1 Rabbit Polyclonal Antibody [clone: S434]
Catalog # BWRLBS4078

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-GABAA R beta1
  • Clonality:
    Polyclonal
  • Clone:
    S434
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human GABAA Rbeta1 around the phosphorylation site of Serine 434.

p-GABAA R?1 (S434) pAb detects endogenous levels of GABAA R?1 protein when phosphorylated at Ser434.

Type: Primary
Antigen: p-GABAA Rbeta1
Clonality: Polyclonal
Clone: S434
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat