Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T161]
BWRLBS4033
- Antibody type:Primary
- Antigen symbol:p-Cdk1/Cdc2
- Clonality:Polyclonal
- Clone:T161
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.
p-Cdk1/Cdc2 (T161) pAb detects endogenous levels of p-Cdk1/Cdc2 protein only when phosphorylated at Thr161.
Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: T161
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat