Anti-TCF-4/12 Rabbit Polyclonal Antibody [clone: S616]
BWRLBS1934
- Antibody type:Primary
- Antigen symbol:TCF-4/12
- Clonality:Polyclonal
- Clone:S616
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 580-630 of Human TCF-4.
TCF-4/12 (S616) pAb detects endogenous levels of TCF-4/12 protein.
Type: Primary
Antigen: TCF-4/12
Clonality: Polyclonal
Clone: S616
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat