Orders LinkContactUsLinkComponent
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
Supplier:   Hach
Nitrogen-nitrate reagent sets, LR
Supplier:  Hach
Certificates
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
2 Options Available Below
Product Details & Documents
For determination of nitrogen-nitrate by the cadmium reduction method.
Product Family Options
Product Information
  • Range
    Chemical method
    For unit
    Sample size
    Type
    Tests per kit
    Pk
    Availability
    Price
  • 705-0132
    Nitrogen nitrate reagent set
    Nitrogen nitrate reagent set
    0,1 - 0,50 mg/l NO₃- N
    Cadmium reduction
    All except DR3000, DR2010, DR2000, PC II
    10 ml
    PP
    100
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 138,00 €
    138,00 €
    1 * 100 ST
       138,00 €
  • HACH2245100
    Nitrogen nitrate reagent set
    Nitrogen nitrate reagent set
    0 - 0,50 mg/l NO₃- N
    Cadmium reduction
    DR3000, DR2000
    25 ml
    PP
    100
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 180,00 €
    180,00 €
    1 * 100 Tests
       180,00 €