Order Entry
Germany
ContactUsLinkComponent
 
(12Z)-9,10-Dihydroxy-12-octadecenoic acid ≥98.0%
 
(12Z)-9,10-Dihydroxy-12-octadecenoic acid ≥98.0%

 

  • Purity:
    ≥98%
  • Formulation:
    A solution in methyl acetate
  • Storage:
    −20 °C
  • Stability:
    1 year from the QC date provided