ContactUsLinkComponent
Extractant for nitrate and nitrogen in soils, calcium sulphate powder
Catalog # HACH1232120
Supplier:  Hach
CAS Number:  
undefined
Extractant for nitrate and nitrogen in soils, calcium sulphate powder
Catalog # HACH1232120
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    20 g
  • Description:
    Extractant for nitrate and nitrogen in soils, calcium sulphate powder
  • Tests per kit:
    50
Specifications
About this item
This calcium sulphate extractant is used for nitrate and nitrogen determination in soils.