Order Entry
Germany
ContactUsLinkComponent
SYBR® Safe Stain
SYBR® Safe Stain
Catalog # EDVO608
Supplier:  EDVOTEK
CAS Number:  
undefined
SYBR® Safe Stain
Catalog # EDVO608
Supplier:  EDVOTEK
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    SYBR® Safe Stain

Specifications

About this item

SYBR® Safe Stain is a DNA stain that fluoresces with a bright green colour when excited with UV light. Like ethidium bromide, SYBR® Safe Stain binds specifically to the DNA double helix.

  • SAFE for the Biotechnology Classroom
  • More sensitive than ethidium bromide
  • Non-mutagenic
  • 10,000 X Concentrate for 750 ml
  • Volume: 80 µl
Caution: Store in dark at room temperature over 25 °C.