Order Entry
Germany
ContactUsLinkComponent
Agarose, white powder, high resolution for molecular biology
Agarose, white powder, high resolution for molecular biology
Catalog # ICNA11AGAR0050
Supplier:  MP Biomedicals
CAS Number:  
undefined
Agarose, white powder, high resolution for molecular biology
Catalog # ICNA11AGAR0050
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Identity Test:
    Passes

Specification Test Results

About this item

 :
Melting Pt:< 67 °C (1.5% gel)
Storage Temperature:Ambient
CAS Number:9012-36-6
MDL Number:MFCD00081294