Indubiose A37
25 g
Catalog # ICNA11AGAI0025
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Standard agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae with an Electroendoosmosis (EEO) value of 0.12 making it an essential component in chemical separation techniques.
- For Electrophoresis: Separation of DNA/RNA fragments from 400 to 8000 bp
- CAS Number:9012-36-6
- Melting Pt:86.5…89.5°C (1.5% gel)
- Melting Pt:86.5-89.5°C (1.5%gel)
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



