Urea ≥99%, white prills, Ultrapure
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:57-13-6
:
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Boiling Pt:135 °C (dec.)
Density:1.335 g/cm³ (20 °C)
:60.06 g/mol
Storage Temperature:Ambient