Glycerine ≥99%, clear colourless viscous liquid cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Boiling Pt:166.1 °C (13.3 hPa) (lit.)
Melting Pt:18.2 °C (lit.)
Density:1.2613 g/cm³ (20 °C) (lit.)
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- pH (1% aq soln):5.0-7.5